missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Dibasic Trihydrate, ≥99%, MP Biomedicals
$76.30 - $351.44
Chemical Identifiers
| CAS | 16788-57-1 |
|---|---|
| Molecular Formula | H7K2O7P |
| Molecular Weight (g/mol) | 228.22 |
| MDL Number | MFCD00149926 |
| InChI Key | XQGPKZUNMMFTAL-UHFFFAOYSA-L |
| Synonym | dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate |
| PubChem CID | 16217523 |
| IUPAC Name | dipotassium trihydrate hydrogen phosphate |
| SMILES | O.O.O.[K+].[K+].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19484580
|
MP Biomedicals Inc
0219484580 |
100 g |
Each for $76.30
|
|
|||||
|
ICN19484590
|
MP Biomedicals Inc
0219484590 |
500 g |
Each for $227.64
|
|
|||||
|
ICN19484591
|
MP Biomedicals Inc
0219484591 |
1 kg |
Each for $351.44
|
|
|||||
Chemical Identifiers
| 16788-57-1 | |
| 228.22 | |
| XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
| 16217523 | |
| O.O.O.[K+].[K+].OP([O-])([O-])=O |
| H7K2O7P | |
| MFCD00149926 | |
| dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
| dipotassium trihydrate hydrogen phosphate |
Specifications
| 16788-57-1 | |
| ≥99% | |
| MFCD00149926 | |
| XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
| dipotassium trihydrate hydrogen phosphate | |
| 16217523 | |
| Potassium Phosphate Dibasic Trihydrate |
| 100 g | |
| H7K2O7P | |
| dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
| O.O.O.[K+].[K+].OP([O-])([O-])=O | |
| 228.22 | |
| 228.2 |
Safety and Handling
Recommended Storage : Room Temperature