Learn More
Potassium hydroxide, 0.5N Standardized Solution in methanol
CAS: 1310-58-3 | HKO | 56.11 g/mol
117,00 $ - 666,78 $
Identifiants chimiques
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| SMILES | [OH-].[K+] |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AA42735K2
|
Thermo Scientific Chemicals
042735K2 |
1 L |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA42735K7
|
Thermo Scientific Chemicals
042735K7 |
4 L |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA42735M4
|
Thermo Scientific Chemicals
042735M4 |
20 L |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Potassium hydroxide, 0.5N standardized solution in methanol is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| [OH-].[K+] |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 |
Spécifications
| Colorless | |
| HKO | |
| UN3286 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| 56.11 | |
| 1 L | |
| Potassium hydroxide Standardized Solution |
| 1310-58-3 | |
| MFCD00003553 | |
| 0.5N in methanol | |
| Miscible with water. | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Like alcohol | |
| Liquid |
Sécurité et manipulation
GHS H Statement
H225-H311-H331-H370-H314
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P310-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H301+H311+H331-H315-H318-H370
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber : 215-181-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only