missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferrocyanide, 10% (w/v), Ricca Chemical
$205.43 - $443.30
Chemical Identifiers
| CAS | 14459-95-1 |
|---|---|
| Molecular Formula | C6H6FeK4N6O3 |
| Molecular Weight (g/mol) | 422.39 |
| MDL Number | MFCD00167023 |
| InChI Key | NLYKFUFIUWPPIZ-UHFFFAOYSA-N |
| IUPAC Name | tetrapotassium hexacyanoironbis(ylium) trihydrate |
| SMILES | O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6096250
|
Ricca Chemical Company
6096250 |
250 mL | Natural Poly Bottle |
Each for $205.43
|
|
||||
|
6096-16
|
Ricca Chemical Company
609616 |
500 mL | Natural Poly Bottle |
Each for $335.93
|
|
||||
|
6096-32
|
Ricca Chemical Company
609632 |
1 L | Natural Poly Bottle |
Each for $443.30
|
|
||||
Chemical Identifiers
| 14459-95-1 | |
| 422.39 | |
| NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
| O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
| C6H6FeK4N6O3 | |
| MFCD00167023 | |
| tetrapotassium hexacyanoironbis(ylium) trihydrate |
Specifications
| Yellow | |
| 89.0,8.91 | |
| C6H6FeK4N6O3 | |
| NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
| tetrapotassium hexacyanoironbis(ylium) trihydrate | |
| 10% | |
| Natural Poly Bottle | |
| 250 mL | |
| 10% (w/v) |
| 14459-95-1 , 7732-18-5 | |
| 93.0,9.28 | |
| MFCD00167023 | |
| O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N | |
| 422.39 | |
| Laboratory | |
| Aqueous Solution | |
| Liquid | |
| Potassium Ferrocyanide |