missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide, 10% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 609616

 View more versions of this product

Catalog No. 6096-16


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Ferrocyanide
14459-95-1
MFCD00167023
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
422.39
Laboratory
Natural Poly Bottle
Labeled with manufacturing date
C6H6FeK4N6O3
NLYKFUFIUWPPIZ-UHFFFAOYSA-N
tetrapotassium hexacyanoironbis(ylium) trihydrate
10%
10% (w/v) Aqueous Solution
500 mL

Chemical Identifiers

14459-95-1
422.39
NLYKFUFIUWPPIZ-UHFFFAOYSA-N
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
C6H6FeK4N6O3
MFCD00167023
tetrapotassium hexacyanoironbis(ylium) trihydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions