missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide, 10% (w/v), Ricca Chemical

Catalog No. 6096250
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6096250 250 mL Natural Poly Bottle
609616 500 mL Natural Poly Bottle
609632 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6096250 Supplier Ricca Chemical Company Supplier No. 6096250

Chemical Identifiers

CAS 14459-95-1 , 7732-18-5
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Yellow
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 250 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Potassium Ferrocyanide
Show More Show Less