missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, 50% Aqueous Solution, MP Biomedicals™

$96.81 - $368.75

Identifiants chimiques

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
ICN19544480
Afficher les documents
MP Biomedicals Inc
0219544480
100 mL
chaque for $96.81
Only null left
 
ICN19544490
missing translation for 'viewDocuments'
MP Biomedicals Inc
0219544490
500 mL
chaque for $291.79
Only null left
 
ICN19544491
missing translation for 'viewDocuments'
MP Biomedicals Inc
0219544491
1 L
chaque for $368.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9002-98-6
43.07
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
((C2H5N)zC2H4N)y(C2H5N)x
MFCD00084427
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
CHEBI:30969
*-CCNCCN(-*)CCN-*

Specifications

Viscous Liquid
((C2H5N)zC2H4N)y(C2H5N)x
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
50,000 to 100,000
9002-98-6
MFCD00084427
100 mL
*-CCNCCN(-*)CCN-*
43.07
CHEBI:30969
Polyethyleneimine
Videos
SDS
Documents

Documents