missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, 50% Aqueous Solution, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219544490

 View more versions of this product

Catalog No. ICN19544490


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Polyethyleneimine
9002-98-6
MFCD00084427
500 mL
*-CCNCCN(-*)CCN-*
43.07
CHEBI:30969
Viscous Liquid
((C2H5N)zC2H4N)y(C2H5N)x
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
50,000 to 100,000

Chemical Identifiers

9002-98-6
43.07
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
((C2H5N)zC2H4N)y(C2H5N)x
MFCD00084427
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
CHEBI:30969
*-CCNCCN(-*)CCN-*
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions