Learn More
Bel-Art™ Polyethylene Utility Bags
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Supplier:  Bel-Art™ H131781012
 Afficher plus de versions de ce produit
Numéro de catalogue. 13-717-281
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Spécifications
Spécifications
Not Autoclavable
Clear
Polyethylene
0.102 mm
10 in.
12 in.
Sample Bag
Zip Tie
Single-use
Non-sterile
10 x 12 in. (254 x 305mm)
254 mm
305 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS