missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polyester Adhesive Warning Sign: NO FLAMMABLE LIQUIDS BEYOND THIS POINT

Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$13.41 - $34.50

Spécifications

Finish Laminated
Material Polyester
Quantity 1 Ea.
Adhesive Type Cold-temperature Permanent Acrylic
Mounting Type Self-adhesive
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Dimensions (L x W) Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Dimensions (L x W) Prix Quantité  
19-105-839
Afficher les documents
Brady™
126344
10 x 7 in.
chaque for $13.41
Il en reste null
 
19-105-842
Afficher les documents
Brady™
126347
14 x 10 in.
chaque for $34.50
Il en reste null
 
Description

Description

  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Spécifications

Spécifications

Laminated
1 Ea.
Self-adhesive
-40°F
Rectangular
NO FLAMMABLE LIQUIDS BEYOND THIS POINT
Polyester
Cold-temperature Permanent Acrylic
230°F
0.01 in.
-40°F to 230°F
Red on White
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'