Learn More
Promega PolyATtract™ mRNA Isolation Systems
The PolyATtract System isolates messenger RNA directly from total RNA.
Supplier: Promega Z5210
Description
- Isolates Highly Pure mRNA from Total RNA
- Protocol complete in approximately 45 minutes
- Isolated mRNA is suitable for use in in vitro translation, RT-PCR and cDNA synthesis
- Cat.# Z5200 and Z5210 contain sufficient reagents for 3 separate mRNA isolations, each from 1-5mg total RNA
- Cat.# Z5300 and Z5310 contain sufficient reagents for 15 separate mRNA isolations, each from 100-1,000μg total RNA
Spécifications
| 2°C to 10°C; Nuclease-free Water 3 x 25mL, SSC (20X) 3 x 1.4mL, Biotinylated Oligo(dT) probe 1 x 35μL, mRNA user tube 3 x 1, Streptavidin MagneSphere™ paramagnetic particles 3 x 3mL | |
| In-vitro translation, RT-PCR and cDNA synthesis | |
| mRNA Isolation System only | |
| 3 Preps |