missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PLK1, His Tag Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Invitrogen™ PV3501

 Afficher plus de versions de ce produit

Numéro de catalogue. PV3501


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

PLK1 is a serine/threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis. Polo-like kinase proteins acts by binding and phosphorylating proteins are that already phosphorylated on a specific motif recognized by the POLO box domains. Phosphorylates BORA, BUB1B/BUBR1, CCNB1, CDC25C, CEP55, ECT2, ERCC6L, FBXO5/EMI1, FOXM1, KIF20A/MKLP2, CENPU, NEDD1, NINL, NPM1, NUDC, PKMYT1/MYT1, KIZ, PPP1R12A/MYPT1, PRC1, RACGAP1/CYK4, SGOL1, STAG2/SA2, TEX14, TOPORS, p73/TP73, TPT1 and WEE1. Plays a key role in centrosome functions and the assembly of bipolar spindles by phosphorylating KIZ, NEDD1 and NINL.
Spécifications

Spécifications

P53350
Kinase Assay
5347
Human PLK1, His Tag
Purified
-80°C, Avoid Freeze/Thaw Cycles
cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg
Plk1
Protein
Human
His-tag
-80°C, Avoid Freeze/Thaw Cycles
Ser⁄Thr Kinases: Other
Z'-LYTE™
polo-like kinase 1
Kinases & Inhibitors
Liquid
Signal Transduction
See Label
20mM HEPES with 0.05% Triton X-100, 50% glycerol, 5mM DTT, 0.5mM EDTA, 150mM NaCl, 0.5mM EGTA and no preservative; pH 7.5
73.3 kDa
7.5
10 μg
RUO
PLK1
PLK Kinase Family
Unconjugated
Recombinant
Full length
Biochemical phospho-specific activity (no antibody)
Dry Ice
PLK1
Insect cells
Recombinant, Full Length
Serine/Threonine Kinases
85% by SDS-PAGE
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions