Learn More
Gibco™ PeproGMP® Human FGF-7 (KGF) Recombinant Protein, PeproTech®
Recombinant Protein
$1228.00 - $1228.00
Spécifications
| Accession Number | P21781 |
|---|---|
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 2252 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
01-672-607
|
Gibco™
GMP1001950UG |
50 μg |
chaque for $1,228.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Spécifications
| P21781 | |
| Lyophilized | |
| 2252 | |
| PeproGMP™ Human FGF-7 (KGF) | |
| RUO | |
| Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
| FGF7 | |
| Protein | |
| Recombinant | |
| -20°Cor to +80°C if preferred | |
| E. coli | |
| ≥ 95% by SDS-PAGE gel and HPLC analyses. |
| Functional Assay | |
| protein with no preservative | |
| 18.9 kDa | |
| HPLC, SDS-PAGE | |
| <0.1 EU/ μg | |
| KGF | |
| Determined by the proliferation of 4 MBr5 cells. | |
| Unconjugated | |
| CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT | |
| Determined by the proliferation of 4 MBr5 cells. | |
| Lyophilized |