Learn More
Thermo Scientific™ Particle-Certified Glass Containers
GREENER_CHOICE
Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
$392.12 - $1,312.88
Specifications
Material Glass
View More Specs
Products 17
Catalog Number Mfr. No. Capacity (Metric) Product Type Color No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Product Type Color No. per Case Price Quantity & Availability  
12200111
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
130 005/LP
15 mL Sample Bottle Amber 57
Case of 57 Each for $1,290.56
Only null left
12200160
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
140 40C/LP
40 mL Container Clear 90
Case of 90 Each for $1,312.88
Only null left
12100001
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
120 02A/LP
60 mL Packer Bottle Amber 24
Case of 24 Each for $772.06
Only null left
12200138
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
114 060A/LP
60 mL Boston Round Bottle Amber 24
Case of 24 Each for $744.09
Only null left
12200101
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
114 125A/LP
125 mL Boston Round Bottle Amber 12
Case of 12 Each for $737.09
Only null left
12200104
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
120 04A/LP
125 mL Packer Bottle Amber 12
Case of 12 Each for $597.12
Only null left
12200102
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
114 125CT/LP
125 mL Boston Round Bottle Clear 12
Case of 12 Each for $466.59
Only null left
12100002
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
121 08A/LP
250 mL Packer Bottle Amber 12
Case of 12 Each for $708.00
Only null left
12200103
View Documents Promotion Details
Thermo Scientific™
114 250A/LP
250 mL Boston Round Bottle Amber 12
Case of 12 Each for $751.09
Only null left
12100000
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
114 250C/LP
250 mL Boston Round Bottle Clear 24
Case of 24 Each for $912.03
Only null left
12200157
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
132 16C/LP
500 mL Sample Jar Clear 12
Case of 12 Each for $741.74
Only null left
12200106
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
122 16A/LP
500 mL Packer Bottle Amber 12
Case of 12 Each for $1,119.62
Only null left
5005728195
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
113500A/LP
500 mL Boston Round Bottle Amber 12
Case of 12 Each for $875.85
Only null left
12200100
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
112 01A/LP
1000 mL Sample Bottle Amber 12
Case of 12 Each for $1,189.59
Only null left
12200159
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
133 32C/LP
1000 mL Sample Jar Clear 12
Case of 12 Each for $392.12
Only null left
12200156
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
110 80A/LP
2500 mL Container Amber 6
Case of 6 Each for $622.79
Only null left
12200137
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
111 04A/LP
4000 mL Jug Amber 4
Case of 4 Each for $595.65
Only null left
Description
Description
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
  • Low particle applications
Specifications
Specifications
Glass