missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Particle-Certified Glass Containers

Description
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Available in clear or amber
- Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
- Individual bottles double bagged to maintain integrity until point of use
- 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
- Low particle applications
Specifications
Specifications
| Capacity | 2 oz., 60 mL |
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Amber |
| Description | Amber Glass Packer Bottle with Lined Closure, 60 mL |
| Diameter (English) | 1.75 in. |
| Material | Glass |
| No. per Case | 24 |
| Height (Metric) | 76.2 mm |
| Product Type | Packer Bottle |
| Show More |