Learn More
Thermo Scientific™ Particle-Certified Glass Containers

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Supplier: Thermo Scientific™ 120 04A/LP
Description
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Available in clear or amber
- Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
- Individual bottles double bagged to maintain integrity until point of use
- 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
- Low particle applications
Chemical Identifiers
| Glass |
Specifications
| 4 oz., 125 mL | |
| 4 oz. | |
| Amber Glass Packer Bottle with PTFE-lined Polypropylene Closure, 125 mL | |
| Glass | |
| 95.25 mm | |
| 3.75 in. | |
| Polypropylene, Low-shedding | |
| 50.8 mm |
| 125 mL | |
| Amber | |
| 2 in. | |
| 12 | |
| Packer Bottle | |
| PTFE | |
| 38-400 mm |