missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Particle-Certified Glass Containers
GREENER_CHOICE

Catalog No. 12100000
Change view
Click to view available options
Capacity (Metric):
15 mL
40 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
2500 mL
4000 mL
Product Type:
Boston Round Bottle
Container
Jug
Packer Bottle
Sample Bottle
Sample Jar
Color:
Amber
Clear
No. per Case:
12
24
4
57
6
90
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Product Type Color No. per Case
12100000 250 mL Boston Round Bottle Clear 24
12200111 15 mL Sample Bottle Amber 57
12200160 40 mL Container Clear 90
12100001 60 mL Packer Bottle Amber 24
12200138 60 mL Boston Round Bottle Amber 24
12200101 125 mL Boston Round Bottle Amber 12
12200104 125 mL Packer Bottle Amber 12
12200102 125 mL Boston Round Bottle Clear 12
12100002 250 mL Packer Bottle Amber 12
12200103 250 mL Boston Round Bottle Amber 12
12200157 500 mL Sample Jar Clear 12
12200106 500 mL Packer Bottle Amber 12
5005728195 500 mL Boston Round Bottle Amber 12
12200100 1000 mL Sample Bottle Amber 12
12200159 1000 mL Sample Jar Clear 12
12200156 2500 mL Container Amber 6
12200137 4000 mL Jug Amber 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 12100000 Supplier Thermo Scientific™ Supplier No. 114 250C/LP
Only null left

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.

Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.

Key Features:

  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging

Recommended for:

  • Low particle applications

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Clear
Description Clear Glass Boston Round Bottle with Closed-top Closure, 250 mL
Diameter (English) 2.375 in.
Material Glass
No. per Case 24
Height (Metric) 136.53 mm
Product Type Boston Round Bottle
Height (English) 5.375 in.
Closure Liner PTFE/Silicone
Closure Material Polypropylene, Low-shedding
Closure Size 24-414 mm
Diameter (Metric) 60.33 mm
Closure Type Closed-top
Show More Show Less