Learn More
Thermo Scientific™ Particle-Certified Glass Containers

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Supplier: Thermo Scientific™ 112 01A/LP
Description
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Available in clear or amber
- Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
- Individual bottles double bagged to maintain integrity until point of use
- 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
- Low particle applications
Chemical Identifiers
| Glass |
Specifications
| 32 oz., 1000 mL | |
| 32 oz. | |
| Amber Glass Narrow Mouth Bottle with Closure, 1000 mL | |
| Glass | |
| Narrow | |
| Sample Bottle | |
| PTFE/Silicone | |
| 33-430 mm |
| 1000 mL | |
| Amber | |
| 3.75 in. | |
| 12 | |
| 215.9 mm | |
| 8.5 in. | |
| Polypropylene, Low-shedding | |
| 95.25 mm |