missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Dioxane, 99.0% min., ACS, MilliporeSigma™
Supplier: Technidata DX20903
Spécifications
| p-Dioxane | |
| 123-91-1 | |
| 99.0%, min. | |
| -O(CH2)2O(CH2)2- | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 1,4-dioxane | |
| 31275 | |
| 88.1 |
| For ACS analysis | |
| 4 L | |
| C4H8O2 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| C1COCCO1 | |
| 88.106 | |
| CHEBI:47032 | |
| ACS/NF/USP |
Identifiants chimiques
| 123-91-1 | |
| 88.106 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| CHEBI:47032 | |
| C1COCCO1 |
| C4H8O2 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 31275 | |
| 1,4-dioxane |