missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Dioxane, 99.0% min., ACS, MilliporeSigma™
Specifications
Specifications
| Quantity | 500 mL |
| Assay Percent Range | 99.0%, min. |
| Linear Formula | -O(CH2)2O(CH2)2- |
| Formula Weight | 88.1 |
| Grade | ACS/NF/USP |
| Chemical Name or Material | p-Dioxane, For ACS analysis |