Learn More
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.
Supplier: Thermo Scientific™ 910710
Description
Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.
- Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
- Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Pouches are 15mL each and are available in 10 packs or 25 packs
Note:
The following part numbers have been changed due to a recent system upgrade.
Old Part Number: 910410
New Part Number: 910410-WA
Old Part Number: 911025
New Part Number: 911025-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Spécifications
| pH Buffer, individual use pouches | |
| Light Yellow | |
| pH calibration | |
| 7 | |
| 10 x 15 mL | |
| Water, Dibasic, Potassium Dihydrogen Phosphate, 5-Bromo-5-Nitro-1,3-Dioxane, FD & C Yellow #5, Sodium Phosphate | |
| Yes |
| 100°C | |
| pH 7.00 Buffer | |
| pH Meter and Electrode, single-use | |
| Liquid | |
| Buffer | |
| Pouch | |
| Soluble |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.