missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ NuTech™ Sleeves

Numéro de catalogue. 19810577
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size
19810577 X-Large
19810578 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 19810577 Fournisseur AlphaProTech Code fournisseur MS016064
Il en reste null

Microporous sleeves are designed with sonic welded seams for added durability and protection.

  • Designed with sonic welded seams for added durability and protection
  • For use in many general-use applications
  • Tapered design with elastic at both ends

Spécifications

Product Type Sleeve Protector
Construction Tapered
Size X-Large
Material Microporous Laminate
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Ultrasonically Welded
Cuff Style Elastic
Latex Free Yes
Model MS-01606
For Use With (Application) Cleanrooms/Controlled Environments
Certifications/Compliance 21C CFR Part 820
Packaging Bulk
No. per Case 300
Cleanroom Class Class 10,000 (ISO 7)
Afficher plus de résultats Afficher moins de résultats