Learn More
AlphaProTech Critical Cover™ NuTech™ Frocks
Numéro de catalogue. p-7113127
Change view
Click to view available options
Size:
2X-Large/3X-Large
4X-Large/5X-Large
Large/X-Large
Small/Medium
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size
19810569 Small/Medium
19810570 Large/X-Large
19810571 2X-Large/3X-Large
19810572 4X-Large/5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 19810569 Fournisseur AlphaProTech Code fournisseur FK621212
Il en reste null
Microporous composite material frock is ideal for visitors and other wearers who do not require highly protective garments.
  • Protects clothes while offering comfort and breathability
  • Elastic wrist allows easy donning of gloves
  • Protective sleeves cut to an appropriate length as safeguard for full coverage
  • Snap collar provides additional coverage of the chest and neck area
Spécifications
Product Type Controlled Environments Frock
Size Small/Medium
Material Microporous Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Seam Type Serged
Collar Type Snap
Sleeve Style Raglan Sleeve
Afficher plus de résultats Afficher moins de résultats