missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, Dilute R (125 g/L HNO3), Ricca Chemical

$276.78 - $276.78

Identifiants chimiques

CAS 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
5330-32
FDS Afficher les documents
Ricca Chemical Company
533032
1 L Amber Glass Bottle
chaque for $276.78
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

7732-18-5
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O

Spécifications

Colorless to Yellow
82.0,15.55
86.0,16.18
1 L
Liquid
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
O[N+]([O-])=O
63.01
CHEBI:48107
Laboratory
7732-18-5,7697-37-2
(125g/L HNO3)
Amber Glass Bottle
HNO3
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
12.55% V/V
Nitric Acid, Dilute Reagent
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'