missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, Dilute R (125 g/L HNO3), Ricca Chemical

Catalog No. 533032
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
533032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 533032 Supplier Ricca Chemical Company Supplier No. 533032
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
Concentration (125g/L HNO3)
CAS Min % 82.0, 15.55
CAS Max % 86.0, 16.18
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
Percent Purity 12.55% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid, Dilute Reagent