missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Boc-D-lysine, 98%
CAS: 31202-69-4 | C11H22N2O4 | 246.31 g/mol
$92.85 - $243.62
Identifiants chimiques
| CAS | 31202-69-4 |
|---|---|
| Molecular Formula | C11H22N2O4 |
| Molecular Weight (g/mol) | 246.31 |
| MDL Number | MFCD00076958 |
| InChI Key | VVQIIIAZJXTLRE-SVGMAFHSNA-N |
| Synonym | h-d-lys boc-oh, r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, n-epsilon-boc-d-lysine, d-lysine, n6-1,1-dimethylethoxy carbonyl, 2r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, ambotzhaa6310, n6-tert-butoxycarbonyl-d-lysine, n-epsilon-t-butyloxycarbonyl-d-lysine |
| PubChem CID | 7018774 |
| IUPAC Name | (2R)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
| SMILES | CC(C)(C)OC(=O)NCCCC[C@@H](N)C(O)=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAH62736MD
|
Thermo Scientific Chemicals
H62736MD |
250 mg |
chaque for $92.85
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAH6273603
|
Thermo Scientific Chemicals
H6273603 |
1 g |
chaque for $243.62
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 31202-69-4 | |
| 246.31 | |
| VVQIIIAZJXTLRE-SVGMAFHSNA-N | |
| 7018774 | |
| CC(C)(C)OC(=O)NCCCC[C@@H](N)C(O)=O |
| C11H22N2O4 | |
| MFCD00076958 | |
| h-d-lys boc-oh, r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, n-epsilon-boc-d-lysine, d-lysine, n6-1,1-dimethylethoxy carbonyl, 2r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, ambotzhaa6310, n6-tert-butoxycarbonyl-d-lysine, n-epsilon-t-butyloxycarbonyl-d-lysine | |
| (2R)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Spécifications
| 31202-69-4 | |
| C11H22N2O4 | |
| h-d-lys boc-oh, r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, n-epsilon-boc-d-lysine, d-lysine, n6-1,1-dimethylethoxy carbonyl, 2r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, ambotzhaa6310, n6-tert-butoxycarbonyl-d-lysine, n-epsilon-t-butyloxycarbonyl-d-lysine | |
| CC(C)(C)OC(=O)NCCCC[C@@H](N)C(O)=O | |
| 250 mg | |
| 7018774 | |
| 98% | |
| Nepsilon-Boc-D-lysine |
| White | |
| MFCD00076958 | |
| VVQIIIAZJXTLRE-SVGMAFHSNA-N | |
| (2R)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 246.31 | |
| 246.3 | |
| Powder |
Sécurité et manipulation
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only