missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Boc-D-lysine, 98%
CAS: 31202-69-4 | C11H22N2O4 | 246.31 g/mol
Supplier: Thermo Scientific Chemicals H62736MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Nepsilon-Boc-D-lysine | |
| White | |
| MFCD00076958 | |
| VVQIIIAZJXTLRE-SVGMAFHSNA-N | |
| (2R)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 246.31 | |
| 246.3 | |
| Powder |
| 31202-69-4 | |
| C11H22N2O4 | |
| h-d-lys boc-oh, r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, n-epsilon-boc-d-lysine, d-lysine, n6-1,1-dimethylethoxy carbonyl, 2r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, ambotzhaa6310, n6-tert-butoxycarbonyl-d-lysine, n-epsilon-t-butyloxycarbonyl-d-lysine | |
| CC(C)(C)OC(=O)NCCCC[C@@H](N)C(O)=O | |
| 250 mg | |
| 7018774 | |
| 98% |
Chemical Identifiers
| 31202-69-4 | |
| 246.31 | |
| VVQIIIAZJXTLRE-SVGMAFHSNA-N | |
| 7018774 | |
| CC(C)(C)OC(=O)NCCCC[C@@H](N)C(O)=O |
| C11H22N2O4 | |
| MFCD00076958 | |
| h-d-lys boc-oh, r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, n-epsilon-boc-d-lysine, d-lysine, n6-1,1-dimethylethoxy carbonyl, 2r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid, ambotzhaa6310, n6-tert-butoxycarbonyl-d-lysine, n-epsilon-t-butyloxycarbonyl-d-lysine | |
| (2R)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only