Learn More
Thermo Scientific™ Nalgene™ Utility Funnels
GREENER_CHOICE
Replace glass lab funnels with these utility funnels, which are an excellent shatter-proof alternative.
$214.20 - $933.39
Produits 3
Numéro de catalogue Numéro du manufacturier. Diameter (Metric)Top Capacity (Metric) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Diameter (Metric)Top Capacity (Metric) Prix Quantity & Availability  
1034810E
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
42560234
62 mm 62 mL
paquet de 12 chaque for $214.20
caisse de 24 paquet for $5,140.80
Il en reste null
1034811A
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
42560414
98 mm 270 mL
paquet de 12 chaque for $262.73
caisse de 12 paquet for $3,152.76
Il en reste null
1043723E
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
42560638
150 mm 1059 mL
paquet de 12 chaque for $933.39
caisse de 6 paquet for $5,600.34
Il en reste null
Description
Description
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.
  • 4256-0234 and 4256-0414 are made from shatter-proof, autoclavable polypropylene.
  • 4256-0638 has a large throat opening for faster transfer of materials and is made from shatter-proof high-density polyethylene (Not autoclavable).
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.   Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and High Density Polyethylene (HDPE) labware at end of product life in communities where these materials can be recycled.  Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Videos
FDS