missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Utility Funnels
GREENER_CHOICE

Catalog No. 1043723E
Change view
Click to view available options
Diameter (Metric)Top:
62 mm
98 mm
150 mm
Capacity (Metric):
62 mL
270 mL
1059 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top Capacity (Metric)
1043723E 150 mm 1059 mL
1034810E 62 mm 62 mL
1034811A 98 mm 270 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1043723E Supplier Thermo Scientific™ Supplier No. 42560638

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Replace glass lab funnels with these utility funnels, which are an excellent shatter-proof alternative.

Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.

  • 4256-0234 and 4256-0414 are made from shatter-proof, autoclavable polypropylene.
  • 4256-0638 has a large throat opening for faster transfer of materials and is made from shatter-proof high-density polyethylene (Not autoclavable).

Specifications

Diameter (Metric)Top 150 mm
Capacity (Metric) 1059 mL
Diameter (Metric) Stem 48 mm
Material HDPE
Height (Metric) 175 mm
Autoclavable Not Autoclavable
Description Funnel Utility HDPE 162mm
Length (Metric) Stem 66 mm
Quantity 12/Pk.; 72/Cs.