Learn More
Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with these strong, durable racks.
$110.89 - $132.79
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Material | RESMER |
| Shape | Rectangular |
| Product Type | Test Tube Rack |
| Catalog Number | Mfr. No. | Color | Array | For Use With (Equipment) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Array | For Use With (Equipment) | Price | Quantity & Availability | |||
1480960
![]() |
Thermo Scientific™
59700513 |
Red | 6 x 12 | 13mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480941
![]() |
Thermo Scientific™
59700313 |
Blue | 6 x 12 | 13mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480945
![]() |
Thermo Scientific™
59700413 |
Green | 6 x 12 | 13mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480937
![]() |
Thermo Scientific™
59700213 |
Yellow | 6 x 12 | 13mm Tubes |
Each for $126.57
Case of 8 Each for $1,012.56
|
|
|||
1480932
![]() |
Thermo Scientific™
59700113 |
Orange | 6 x 12 | 13mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480922
![]() |
Thermo Scientific™
59700013 |
White | 6 x 12 | 13mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480962
![]() |
Thermo Scientific™
59700516 |
Red | 6 x 12 | 16mm Tubes |
Each for $110.89
Case of 8 Each for $887.12
|
|
|||
1480943
![]() |
Thermo Scientific™
59700316 |
Blue | 6 x 12 | 16mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480947
![]() |
Thermo Scientific™
59700416 |
Green | 6 x 12 | 16mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480939
![]() |
Thermo Scientific™
59700216 |
Yellow | 6 x 12 | 16mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480935
![]() |
Thermo Scientific™
59700116 |
Orange | 6 x 12 | 16mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480924
![]() |
Thermo Scientific™
59700016 |
White | 6 x 12 | 16mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480964
![]() |
Thermo Scientific™
59700520 |
Red | 4 x 10 | 20mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480952
![]() |
Thermo Scientific™
59700320 |
Blue | 4 x 10 | 20mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480946
![]() |
Thermo Scientific™
59700420 |
Green | 4 x 10 | 20mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480938
![]() |
Thermo Scientific™
59700220 |
Yellow | 4 x 10 | 20mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480931
![]() |
Thermo Scientific™
59700120 |
Orange | 4 x 10 | 20mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480926
![]() |
Thermo Scientific™
59700020 |
White | 4 x 10 | 20mm Tubes |
Each for $118.89
Case of 8 Each for $951.12
|
|
|||
1480966
![]() |
Thermo Scientific™
59700525 |
Red | 4 x 10 | 25mm Tubes |
Each for $132.04
Case of 8 Each for $1,056.32
|
|
|||
1480942
![]() |
Thermo Scientific™
59700325 |
Blue | 4 x 10 | 25mm Tubes |
Each for $128.04
Case of 8 Each for $1,024.32
|
|
|||
1480948
![]() |
Thermo Scientific™
59700425 |
Green | 4 x 10 | 25mm Tubes |
Each for $132.79
Case of 8 Each for $1,062.32
|
|
|||
1480940
![]() |
Thermo Scientific™
59700225 |
Yellow | 4 x 10 | 25mm Tubes |
Each for $132.79
Case of 8 Each for $1,062.32
|
|
|||
1480933
![]() |
Thermo Scientific™
59700125 |
Orange | 4 x 10 | 25mm Tubes |
Each for $123.86
Case of 8 Each for $990.88
|
|
|||
1480928
![]() |
Thermo Scientific™
59700025 |
White | 4 x 10 | 25mm Tubes |
Each for $128.04
Case of 8 Each for $1,024.32
|
|
|||
1480968
![]() |
Thermo Scientific™
59700530 |
Red | 3 x 8 | 30mm Tubes |
Each for $128.04
Case of 8 Each for $1,024.32
|
|
|||
1480944
![]() |
Thermo Scientific™
59700330 |
Blue | 3 x 8 | 30mm Tubes |
Each for $128.04
Case of 8 Each for $1,024.32
|
|
|||
1480949
![]() |
Thermo Scientific™
59700430 |
Green | 3 x 8 | 30mm Tubes |
Each for $129.86
Case of 8 Each for $1,038.88
|
|
|||
1480950
![]() |
Thermo Scientific™
59700230 |
Yellow | 3 x 8 | 30mm Tubes |
Each for $129.86
Case of 8 Each for $1,038.88
|
|
|||
1480936
![]() |
Thermo Scientific™
59700130 |
Orange | 3 x 8 | 30mm Tubes |
Each for $129.86
Case of 8 Each for $1,038.88
|
|
|||
1480930
![]() |
Thermo Scientific™
59700030 |
White | 3 x 8 | 30mm Tubes |
Each for $128.04
Case of 8 Each for $1,024.32
|
|
|||
Description
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks. Molded in a single piece for exceptional strength and durability, these racks have no coating to chip, peel or rust.
- Smooth finish leaves fewer places for contaminants to lodge; no welds to break, no sharp edges.
- Endplates accept large labels for easy identification of samples.
- Higher specific gravity than water so it stays submerged in a water bath and won't float.
- Withstands most lab chemicals and temperatures up to 121°C for a variety of lab procedures.
- Available in six bright colors to help keep samples organized.
- 15mL conical tubes fit 20mm racks; 50mL conical fit 30mm racks.
- No assembly is required; rack is ready to use.
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
| Autoclavable | |
| RESMER | |
| Rectangular | |
| Test Tube Rack |