missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine 98.0+%, TCI America™
Supplier: TCI America B53505G
Specifications
| N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine | |
| 95°C | |
| MFCD00065604 | |
| boc-d-tyr me-oh, boc-4-methoxy-d-phenylalanine, boc-o-methyl-d-tyrosine, boc-4-methoxy-d-phe-oh, n-boc-4-methoxy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid, d-tyrosine,n-1,1-dimethylethoxy carbonyl-o-methyl, d-tyrosine, n-1,1-dimethylethoxy carbonyl-o-methyl, boc-l-phe 4-och3-oh, boc-d-phe 4-ome-oh | |
| CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)OC)C(=O)O | |
| 295.335 | |
| 295.34 |
| 68856-96-2 | |
| C15H21NO5 | |
| 5 g | |
| SLWWWZWJISHVOU-GFCCVEGCSA-N | |
| (2R)-3-(4-methoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 7017908 | |
| ≥98.0% (T) |
Chemical Identifiers
| 68856-96-2 | |
| 295.335 | |
| SLWWWZWJISHVOU-GFCCVEGCSA-N | |
| 7017908 | |
| CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)OC)C(=O)O |
| C15H21NO5 | |
| MFCD00065604 | |
| boc-d-tyr me-oh, boc-4-methoxy-d-phenylalanine, boc-o-methyl-d-tyrosine, boc-4-methoxy-d-phe-oh, n-boc-4-methoxy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid, d-tyrosine,n-1,1-dimethylethoxy carbonyl-o-methyl, d-tyrosine, n-1,1-dimethylethoxy carbonyl-o-methyl, boc-l-phe 4-och3-oh, boc-d-phe 4-ome-oh | |
| (2R)-3-(4-methoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Safety and Handling
TSCA : No