missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine 98.0+%, TCI America™

Catalog No. p-7124051
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53501G 1 g
B53505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53501G Supplier TCI America Supplier No. B53501G
Only null left

Chemical Identifiers

CAS 68856-96-2
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00065604
InChI Key SLWWWZWJISHVOU-GFCCVEGCSA-N
Synonym boc-d-tyr me-oh, boc-4-methoxy-d-phenylalanine, boc-o-methyl-d-tyrosine, boc-4-methoxy-d-phe-oh, n-boc-4-methoxy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid, d-tyrosine,n-1,1-dimethylethoxy carbonyl-o-methyl, d-tyrosine, n-1,1-dimethylethoxy carbonyl-o-methyl, boc-l-phe 4-och3-oh, boc-d-phe 4-ome-oh
PubChem CID 7017908
IUPAC Name (2R)-3-(4-methoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)OC)C(=O)O

Specifications

Melting Point 95°C
Quantity 1 g
Formula Weight 295.34
Percent Purity ≥98.0% (T)
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine