missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl S-Acetylthioglycolate 94.0+%, TCI America™
Supplier: TCI America S04311G
Spécifications
| N-Succinimidyl S-Acetylthioglycolate [Cross-linking Reagent] | |
| 97°C | |
| C8H9NO5S | |
| 1 g | |
| FLCQLSRLQIPNLM-UHFFFAOYSA-N | |
| (2,5-dioxopyrrolidin-1-yl) 2-acetylsulfanylacetate | |
| 127532 | |
| ≥94.0% (GC) |
| 76931-93-6 | |
| White-Yellow | |
| MFCD00036891 | |
| sata, n-succinimidyl-s-acetylthioacetate, 2,5-dioxopyrrolidin-1-yl 2-acetylthio acetate, n-succinimidyl s-acetylthioglycolate, n-hydroxysuccinimide s-acetylthioacetate, 2,5-dioxopyrrolidin-1-yl 2-acetylsulfanylacetate, s-acetylthioglycolic acid n-hydroxysuccinimide ester, hsata, n-succinimidyl s-acetylthioacetate, ethanethioic acid, s-2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl ester | |
| CC(=O)SCC(=O)ON1C(=O)CCC1=O | |
| 231.222 | |
| 231.22 | |
| Crystalline Powder |
Identifiants chimiques
| 76931-93-6 | |
| 231.222 | |
| FLCQLSRLQIPNLM-UHFFFAOYSA-N | |
| 127532 | |
| CC(=O)SCC(=O)ON1C(=O)CCC1=O |
| C8H9NO5S | |
| MFCD00036891 | |
| sata, n-succinimidyl-s-acetylthioacetate, 2,5-dioxopyrrolidin-1-yl 2-acetylthio acetate, n-succinimidyl s-acetylthioglycolate, n-hydroxysuccinimide s-acetylthioacetate, 2,5-dioxopyrrolidin-1-yl 2-acetylsulfanylacetate, s-acetylthioglycolic acid n-hydroxysuccinimide ester, hsata, n-succinimidyl s-acetylthioacetate, ethanethioic acid, s-2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl ester | |
| (2,5-dioxopyrrolidin-1-yl) 2-acetylsulfanylacetate |
Sécurité et manipulation
missing translation for 'tsca' : No
Recommended Storage : Freezer