missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl S-Acetylthioglycolate 94.0+%, TCI America™

Catalog No. p-7135885
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04311G 1 g
S04315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04311G Supplier TCI America Supplier No. S04311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76931-93-6
Molecular Formula C8H9NO5S
Molecular Weight (g/mol) 231.222
MDL Number MFCD00036891
InChI Key FLCQLSRLQIPNLM-UHFFFAOYSA-N
Synonym sata, n-succinimidyl-s-acetylthioacetate, 2,5-dioxopyrrolidin-1-yl 2-acetylthio acetate, n-succinimidyl s-acetylthioglycolate, n-hydroxysuccinimide s-acetylthioacetate, 2,5-dioxopyrrolidin-1-yl 2-acetylsulfanylacetate, s-acetylthioglycolic acid n-hydroxysuccinimide ester, hsata, n-succinimidyl s-acetylthioacetate, ethanethioic acid, s-2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl ester
PubChem CID 127532
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-acetylsulfanylacetate
SMILES CC(=O)SCC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 231.22
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl S-Acetylthioglycolate [Cross-linking Reagent]
TSCA No
Recommended Storage Freezer