missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Isopropylacrylamide, 99%, pure, stabilized

CAS: 2210-25-5 | C6H11NO | 113.16 g/mol

Supplier:  Thermo Scientific Chemicals 412785000

 Afficher plus de versions de ce produit

Numéro de catalogue. AC412785000


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

N-Isopropylacrylamide
2210-25-5
100.0
White to Yellow
Authentic
Plastic bottle
CH2CHCONHCH(CH3)2
500 g
Solubility in water: soluble
CC(C)NC(=O)C=C
113.16
16637
99%
Crystalline Powder or Crystals
99%
98.5
62.0°C to 66.0°C
89.0°C to 92.0°C (2.0 mmHg)
98.5% min. (GC)
C6H11NO
MFCD00041913
n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u
QNILTEGFHQSKFF-UHFFFAOYSA-N
N-propan-2-ylprop-2-enamide
500ppm (ca.) MEHQ
113.16
Pure

Identifiants chimiques

2210-25-5
113.16
QNILTEGFHQSKFF-UHFFFAOYSA-N
16637
CC(C)NC(=O)C=C
C6H11NO
MFCD00041913
n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u
N-propan-2-ylprop-2-enamide
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 218-638-5

missing translation for 'rtecsNumber' : AS3675000

missing translation for 'tsca' : TSCA

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only