Learn More
Thermo Scientific Chemicals N-Isopropylacrylamide, 99%, pure, stabilized
CAS: 2210-25-5 | C6H11NO | 113.16 g/mol
$171.53 - $1497.39
Chemical Identifiers
| CAS | 2210-25-5 |
|---|---|
| Molecular Formula | C6H11NO |
| Molecular Weight (g/mol) | 113.16 |
| MDL Number | MFCD00041913 |
| InChI Key | QNILTEGFHQSKFF-UHFFFAOYSA-N |
| Synonym | n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u |
| PubChem CID | 16637 |
| IUPAC Name | N-propan-2-ylprop-2-enamide |
| SMILES | CC(C)NC(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC412780250
|
Thermo Scientific Chemicals
412780250 |
25 g |
Each for $171.53
|
|
|||||
|
AC412781000
|
Thermo Scientific Chemicals
412781000 |
100 g |
Each for $495.42
|
|
|||||
|
AC412785000
|
Thermo Scientific Chemicals
412785000 |
500 g |
Each for $1,497.39
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2210-25-5 | |
| 113.16 | |
| QNILTEGFHQSKFF-UHFFFAOYSA-N | |
| 16637 | |
| CC(C)NC(=O)C=C |
| C6H11NO | |
| MFCD00041913 | |
| n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u | |
| N-propan-2-ylprop-2-enamide |
Specifications
| 2210-25-5 | |
| 100.0 | |
| White to Yellow | |
| Authentic | |
| Glass Bottle | |
| CH2CHCONHCH(CH3)2 | |
| 25 g | |
| Solubility in water: soluble | |
| CC(C)NC(=O)C=C | |
| 113.16 | |
| 16637 | |
| 99% | |
| Crystalline Powder or Crystals |
| 98.5 | |
| 62.0°C to 66.0°C | |
| 89.0°C to 92.0°C (2.0 mmHg) | |
| 98.5% min. (GC) | |
| C6H11NO | |
| MFCD00041913 | |
| n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u | |
| QNILTEGFHQSKFF-UHFFFAOYSA-N | |
| N-propan-2-ylprop-2-enamide | |
| 500ppm (ca.) MEHQ | |
| 113.16 | |
| Pure | |
| N-Isopropylacrylamide, 99% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 218-638-5
RTECSNumber : AS3675000
TSCA : TSCA
RUO – Research Use Only