missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-N-methyl-D-phenylalanine, 98%
CAS: 85466-66-6 | C15H21NO4 | 279.34 g/mol
97,97 $ - 831,45 $
Identifiants chimiques
| CAS | 85466-66-6 |
|---|---|
| Molecular Formula | C15H21NO4 |
| Molecular Weight (g/mol) | 279.34 |
| MDL Number | MFCD00151890 |
| InChI Key | AJGJINVEYVTDNH-UHFFFAOYNA-N |
| Synonym | boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid |
| PubChem CID | 7015127 |
| SMILES | CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAH62638MD
|
Thermo Scientific Chemicals
H62638MD |
250 mg |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAH6263803
|
Thermo Scientific Chemicals
H6263803 |
1 g |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAH6263806
|
Thermo Scientific Chemicals
H6263806 |
5 g |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 85466-66-6 | |
| 279.34 | |
| AJGJINVEYVTDNH-UHFFFAOYNA-N | |
| 7015127 |
| C15H21NO4 | |
| MFCD00151890 | |
| boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid | |
| CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C |
Spécifications
| 85466-66-6 | |
| C15H21NO4 | |
| boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid | |
| CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C | |
| 250 mg | |
| 7015127 | |
| 98% | |
| N-Boc-N-methyl-D-phenylalanine |
| White | |
| MFCD00151890 | |
| AJGJINVEYVTDNH-UHFFFAOYNA-N | |
| (2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-phenylpropanoic acid | |
| 279.34 | |
| 279.33 | |
| Powder |
Sécurité et manipulation
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only