missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-N-methyl-D-phenylalanine, 98%
CAS: 85466-66-6 | C15H21NO4 | 279.34 g/mol
Supplier: Thermo Scientific Chemicals H6263803
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Boc-N-methyl-D-phenylalanine | |
| White | |
| MFCD00151890 | |
| AJGJINVEYVTDNH-UHFFFAOYNA-N | |
| 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid | |
| 279.34 | |
| 279.33 | |
| Powder |
| 85466-66-6 | |
| C15H21NO4 | |
| boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid | |
| CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C | |
| 1 g | |
| 7015127 | |
| 98% |
Chemical Identifiers
| 85466-66-6 | |
| 279.34 | |
| AJGJINVEYVTDNH-UHFFFAOYNA-N | |
| 7015127 |
| C15H21NO4 | |
| MFCD00151890 | |
| boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid | |
| CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only