missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alanine, 98+%

CAS: 15761-38-3 | C8H15NO4 | 189.21 g/mol

Supplier:  Thermo Scientific Chemicals A1601814

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1601814


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

N-Boc-L-alanine
80°C to 84°C
MFCD00037225
1726365
QVHJQCGUWFKTSE-YFKPBYRVSA-N
C[C@H](NC(=O)OC(C)(C)C)C(O)=O
189.21
189.21
15761-38-3
C8H15NO4
25 g
boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
−26° (c=2 in Acetic Acid)
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
85082
≥98%

Identifiants chimiques

15761-38-3
189.21
QVHJQCGUWFKTSE-YFKPBYRVSA-N
85082
C[C@H](NC(=O)OC(C)(C)C)C(O)=O
C8H15NO4
MFCD00037225
boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 239-847-8

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only