missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alanine, 98+%

Catalog No. p-7023000
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1601806 5 g
AAA1601814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1601806 Supplier Thermo Scientific Chemicals Supplier No. A1601806
Only null left

CAS: 15761-38-3 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 15761-38-3
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037225
InChI Key QVHJQCGUWFKTSE-YFKPBYRVSA-N
Synonym boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
PubChem CID 85082
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES C[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 80°C to 84°C
Quantity 5 g
Beilstein 1726365
Optical Rotation −26° (c=2 in Acetic Acid)
Formula Weight 189.21
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-alanine
EINECSNumber 239-847-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only