missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114114
Description
Immunogen sequence: PMLQTVAKNK DQGTYEDYVE GLRVFDKEGN GTVMGAEIRH VLVTLGEKMT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 86%.
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain that is expressed in smooth muscle and non-muscle tissues. Genomic sequences representing several pseudogenes have been described and two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq, Jul 2008].Spécifications
| MYL6 | |
| Polyclonal | |
| Unconjugated | |
| MYL6 | |
| 17 kDa myosin light chain; ESMLC; LC17; LC17A; LC17B; LC17-GI; LC17-NM; MLC1SM; MLC-3; MLC3NM; MLC3SM; Myl6; Myln; myosin light chain 3; myosin light chain 6; myosin light chain A3; Myosin light chain alkali 3; myosin light chain, alkali, nonmuscle; myosin light polypeptide 6; myosin, light chain 6, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6, alkali, smooth muscle and non-muscle; smooth muscle and nonmuscle myosin light chain alkali 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17904, 4637, 685867 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P60660, Q60605, Q64119 | |
| MYL6 | |
| Synthetic peptide directed towards the middle region of human MYL6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |