Learn More
Contec Multi Bucket System with String Mops
Contec's Multi-Bucket System offers flexibility with two or three buckets in stainless steel or polypropylene.
Supplier:  Contec 2650U2PS
 Afficher plus de versions de ce produit
Numéro de catalogue. 19-991-015
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
The Contec Multi-Bucket System is designed for use with Contec's string mops, offering flexibility and efficiency in cleanroom cleaning. With customizable bucket and wringer options, it enhances convenience, mobility, and organization.
  • Configurable with two or three buckets in stainless steel or polypropylene
  • Extendable front frame accommodates an additional bucket for added capacity
  • Quiet, locking casters for smooth movement and stability
  • Utility handle for easy transport of cleaning supplies
Integrated mop holders with mop brace for secure storage during transport
  • Compatible with stainless steel downpress wringer for string mop use
  • Autoclavable at 121°C for 30 minutes for sterilization in critical environments
Spécifications
Spécifications
2 Bucket System
Polypropylene Bucket, Stainless Steel Cart
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS