Learn More
Contec Multi Bucket System with String Mops
Contec's Multi-Bucket System offers flexibility with two or three buckets in stainless steel or polypropylene.
$4,761.70 - $7,909.17
| Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
|
19991015
|
Contec
2650U2PS |
Polypropylene Bucket, Stainless Steel Cart |
Case for $4,761.70
|
|
|||||
|
19991011
|
Contec
2707U2SS |
Stainless Steel |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The Contec Multi-Bucket System is designed for use with Contec's string mops, offering flexibility and efficiency in cleanroom cleaning. With customizable bucket and wringer options, it enhances convenience, mobility, and organization.
- Configurable with two or three buckets in stainless steel or polypropylene
- Extendable front frame accommodates an additional bucket for added capacity
- Quiet, locking casters for smooth movement and stability
- Utility handle for easy transport of cleaning supplies
Integrated mop holders with mop brace for secure storage during transport
- Compatible with stainless steel downpress wringer for string mop use
- Autoclavable at 121°C for 30 minutes for sterilization in critical environments
Specifications
| 2 Bucket System |