missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Mouse anti-beta-Actin, Clone: 4C2,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABT825
Description
Specifically detects beta-Actin clone: 4C2 in Chicken, Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Western Blotting
Actin, cytoplasmic 1 (UniProt P60709; also known as Beta-actin) is encoded by the ACTB gene (Gene ID 60) in human. Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin is encoded by the ACTA1 gene, smooth muscle alpha-actin by the ACTA2 gene, cytoplasmic beta-actin by the ACTB gene, cardiac muscle alpha-actin by the ACTC1 gene, cytoplasmic gamma-actin by the ACTG1 gene, and smooth muscle gamma-actin by the ACTG2 (a.k.a. ACTA3) gene. Although actins show 90% overall sequence homology, isoforms do show spatial, temporal, and tissue-specific expression patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic β and γ-actins, are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt.Spécifications
| beta-Actin | |
| Monoclonal | |
| Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| ACTB | |
| KLH-conjugated linear peptide corresponding to the the N-terminal sequence of human beta-Actin. | |
| 100 μg | |
| Cell Structure | |
| NP_001092.1 | |
| Stable for one year at 2°C to 8°C from date of receipt. | |
| IgG1 κ |
| Immunocytochemistry, Western Blot | |
| 4C2 | |
| P60709 | |
| Mouse | |
| Protein G Purified | |
| RUO | |
| Primary | |
| Chicken, Human, Mouse, Rat | |
| Purified |
For Research Use Only