Learn More
Invitrogen™ Click-iT™ EdU Alexa Fluor™ 555 HCS Assay
Superior alternative to traditional proliferation assays
$1100.00 - $4560.00
Spécifications
| Color | Orange |
|---|---|
| Detection Method | Fluorescence |
| For Use With (Application) | High-content Screening (HCS) |
| For Use With (Equipment) | High Content Analysis Instrument |
| Product Type | HCS Assay |
Comprend: The kit contains sufficient material for two 96-well plates.
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
C10353
|
Invitrogen™
C10353 |
10 x 96 well plates |
chaque for $4,560.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
C10352
|
Invitrogen™
C10352 |
2 x 96 well plates |
chaque for $1,100.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
The Click-iT EdU Alexa Fluor 555 HCS Assay is a superior alternative to traditional proliferation assays that is optimized for high-content imaging applications. In this assay the modified thymidine analogue EdU is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor dye in a fast, highly specific click reaction. This fluorescent labeling of proliferating cells is accurate and compatible with antibody methods due to the mild click protocol. The most accurate proliferation-detection methods are based on the incorporation and measurement of nucleoside analogues in newly synthesized DNA, with bromodeoxyuridine (BrdU) a commonly used analogue. BrdU-labeled DNA is quantitated using anti-BrdU antibodies following DNA denaturation by harsh methods (HCl, heat, or enzymes) to expose the BrdU molecules. This step is time consuming and difficult to perform consistently. The harsh treatment can also adversely affect sample integrity and quality, which makes co-staining with other antibodies challenging.
- Simple: works the first time, every time, in less time
- Efficient: no denaturation steps or harsh treatment required
- Gentle on samples: better preservation of cell morphology, antigen structure, and DNA integrity
- Consistent: not dependent on variable antibody lots for detection
- Less hazardous
Superior Proliferation Methodology
The Click-iT EdU Alexa Fluor 555 HCS Assay provides a superior alternative to BrdU assays for measuring cell proliferation. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog of thymidine and is incorporated into DNA during active DNA synthesis. With Click-iT EdU, mild fixation and detergent permeabilization is sufficient for the small molecule-based Click-iT EdU detection reagent to gain access to the DNA. As a consequence, the Click-iT EdU HCS assay is not only easy to use, but more accurate and compatible with cell cycle analysis and other intracellular or extracellular targets for truly content-rich results.
Cell Analysis, Cell Cycle, Cell Proliferation, Cell Proliferation with Click Chemistry, Cell Viability, Proliferation and Function, Cellular Imaging, Click Chemistries, DNA Replication and Repair, High-Content Screening (HCS), Labeling Chemistry
Order Info
Shipping Condition: Room temperature
Spécifications
| Orange | |
| High-content Screening (HCS) | |
| HCS Assay | |
| Visible | |
| Less hazardous | |
| Room Temperature |
| Fluorescence | |
| High Content Analysis Instrument | |
| Alexa Fluor™ 555 | |
| 96-well plate | |
| Alexa Fluor, Click-iT |
For Research Use Only. Not for use in diagnostic procedures.