missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Calibration Standards A For Environmental Analysis, Set of 5, SPEX CertiPrep™
Mixed Calibration Multi-Element Standards with Mercury for AA & ICP, 125 mL, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Supplier: SPEX CertiPrep Group LLC MIXSTDSETA
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Mixed Calibration Standards
- 125 mL Volume
- Includes Mercury
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Mixed Calibration Standards | |
Triple-leached LDPE bottle | |
AA, ICP | |
Certified Reference Material | |
Environmental |
Set of 5 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
5 x 125 mL | |
Set contains one 100mL bottle each of:, MIXSTD1A-100, MIXSTD2A-100, MIXSTD3A-100, MIXSTD4A-100, MIXSTD5A-100 |
Product intended for laboratory use only