missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Calibration Standard 4 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC MIXSTD4100
Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ca | 1000μg/mL |
| K | 400μg/mL |
| Al | 200μg/mL |
| Na | 200μg/mL |
| Cr | 20μg/mL |
| Ni | 20μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
| Mixed Calibration Standard 4 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| 5% HNO3 | |
| Environmental |
| Triple-leached LDPE bottle | |
| AA, ICP | |
| Certified Reference Material | |
| 125 mL | |
| Components in 5% HNO3: Al •200μg/mL Ca •1000μg/mL Cr •20μg/mL Ni •20μg/mL K •400μg/mL Na •200μg/mL |
Product intended for laboratory use only