missing translation for 'onlineSavingsMsg'
Learn More

Mixed Calibration Standard 3A For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$773.91 - $773.91

Spécifications

Chemical Name or Material Mixed Calibration Standard 3A
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
voir les spécifications

Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
MIXSTD3A500
Afficher les documents
SPEX CertiPrep Group LLC
MIXSTD3A500
500 mL
chaque for $773.91
Il en reste null
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
P100μg/mL
Ce20μg/mL
Co20μg/mL
V20μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Spécifications

Spécifications

Mixed Calibration Standard 3A
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
5% HNO3
Components in 5% HNO3:
Co    •20μg/mL
P    •100μg/mL
V    •20μg/mL
Triple-leached LDPE bottle
AA, ICP
Certified Reference Material
Environmental
Videos
FDS
Documents

Documents

Product intended for laboratory use only