missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Calibration Standard 1A For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$800.03 - $800.03
Spécifications
| Chemical Name or Material | Mixed Calibration Standard 1A |
|---|---|
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
MIXSTD1A100
|
SPEX CertiPrep Group LLC
MIXSTD1A100 |
125 mL |
chaque for $800.03
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| As | 100μg/mL |
| Ca | 100μg/mL |
| Sb | 50μg/mL |
| Se | 50μg/mL |
| B | 20μg/mL |
| Cd | 20μg/mL |
| Cu | 20μg/mL |
| Mn | 20μg/mL |
| Ba | 10μg/mL |
| Ag | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
| Mixed Calibration Standard 1A | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride | |
| Components in 5% HNO3: Sb •50μg/mL As •100μg/mL Ba •10μg/mL B •10μg/mL Cd •20μg/mL Ca •100μg/mL Cu •20μg/mL Mn •20μg/mL Se •50μg/mL Ag •5μg/mL |
| Triple-leached LDPE bottle | |
| AA, ICP | |
| Certified Reference Material | |
| Environmental |
Product intended for laboratory use only