missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Milk Smear Stain, Ricca Chemical

Modified Newman-Lampert Formulation, for Microscopic Bacterial Examination of Milk

$558.33 - $806.27

Specifications

Chemical Name or Material Methylene Blue Milk Smear Stain
Name Note Modified Newman-Lampert Formulation
CAS 79-34-5 , 64-17-5 , 64-19-7 , 7732-18-5 , 65-56-1 , 7220-79-3
CAS Min % 59.0,33.18,3.53,2.18,0.5
CAS Max % 61.0,34.53,3.67,2.27,0.52
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
4890-16
View Documents
Ricca Chemical Company
489016
500 mL Amber Glass Bottle
Each for $558.33
Only null left
 
4890-32
View Documents
Ricca Chemical Company
489032
1 L Amber Glass Bottle
Each for $806.27
Only null left
 
Description

Description

Specifications

Specifications

Methylene Blue Milk Smear Stain
79-34-5 , 64-17-5 , 64-19-7 , 7732-18-5 , 65-56-1 , 7220-79-3
61.0,34.53,3.67,2.27,0.52
Amber Glass Bottle
Liquid
Modified Newman-Lampert Formulation
59.0,33.18,3.53,2.18,0.5
Laboratory
Dark Blue
Videos
SDS
Documents

Documents