accessibility menu, dialog, popup

UserName

H2OH2OH2OClH3CNCH3NSNCH3CH3

CAS RN 7220-79-3

IUPAC Name: 3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride
Synonyms: 3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride
Molecular Weight (g/mol): 373.9
Molecular Formula: C16H24ClN3O3S
InChi Key: XQAXGZLFSSPBMK-UHFFFAOYSA-M
SMILES: O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
Quantity 
  • (3)
  • (2)
  • (1)
  • (7)
  • (3)
  • (1)
  • (1)
  • (9)
  • (1)
  • (2)
  • (3)
  • (3)
Color 
  • (7)
  • (11)
  • (1)
  • (11)
Grade 
  • (3)
  • (2)
  • (2)
  • (4)
  • (6)
  • (9)
Molecular Weight (g/mol) 
  • (3)
  • (16)
Percent Purity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (8)
  • (3)
  • (2)
  • (3)
  • (1)
  • (10)
  • (8)
  • (9)
  • (3)
  • (2)
  • (1)
  • (7)
  • (3)
  • (1)
  • (1)
  • (9)
  • (1)
  • (2)
  • (3)
  • (3)
  • (7)
  • (11)
  • (1)
  • (11)
  • (3)
  • (2)
  • (2)
  • (4)
  • (6)
  • (9)
  • (3)
  • (16)
  • (1)
  • (2)
  • (3)
  • (3)